Fisherbrand™ Flexible-Arm™ Light
Three-point articulating arm allows lamp to be moved 360° and adjusted to any position
Manufacturer:  Fisherbrand™ CON3095
 View more versions of this product
Catalog No. 15-077-65
Description
Description
Three-point articulating arm allows lamp to be moved 360° and adjusted to any position
Designed for inspection, sample examination and electronics quality control.
  • Finger control pivots, raises and lowers lamp
  • Light (35w halogen) closely resembles daylight for correct color detection
  • Spring counterbalance for easy movement
  • Weighted metal base for stability
Specifications
Specifications
Halogen white
For inspection, sample examination and electronics quality control
20 in.
115V
Flexible Arm Natural Light Lamp
UL
White reflector, 35w soft white Halogen light, 360° three point articulating arm, fingertip control, spring counterbalance, weighted metal base
50.8cm
35w
Documents