Please login to your online account to display your discounted pricing

Fisher Scientific™ Flask Clamps for Incubating Mini Shakers


Available in stainless steel and PVC formats

Manufacturer: troemner llc  980FIPFC12

 View more versions of this product

Catalog No. 02-217-705

  • / Each
 

Description & Specifications

Specifications

Capacity (English) 4.23 oz.
For Use With Incubating mini shaker
Type Clamp
Holds 125mL Flask
Material PVC
Capacity 4.23 oz., 125mL
Capacity (Metric) 125mL