Fisherbrand™ Filling Solutions for accumet™ Double Junction Reference Electrodes
For use with accumet™ Double Junction Reference Electrodes.
Manufacturer:  Fisherbrand™ ORI13620434
 View more versions of this product
Catalog No. 13-620-434
Description
Description
Five 60mL bottles
Specifications
Specifications
Five 60mL bottles
accumet Double Junction Reference Electrode Outer Chamber
Filling Solution
Double
Documents