Please login to your online account to display your discounted pricing
Fisher Scientific™ FH120DH Dispensing Handle
Offers contamination-free operation
$564.77 - $564.77
Specifications
Description Dispensing handle
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Plug Type Price Quantity    
13-310-666
fisher scientific™
7777780
FH100 or FH100X 9-pin Each for $564.77
13-310-667
fisher scientific™
7777790
FH100D or FH100DX 25-pin Each for $564.77
Description & Specifications
Specifications
Description Dispensing handle
Ideal for automated process applications.
Ease of use
  • Contamination free operation
  • Adaptability with various pump models and tubing sizes
  • Intuitive controlsdash (em)Start/Stop, Dispense/Purge and Prime
Ease of maintenance
  • IP31 enclosure rating allows for simple wipe down clean up
  • Tubing runs the full length of the handledash (em)no interior surfaces to clean
Diverse performance range
  • Accommodates a wide range of tubing sizes from 0.8 to 8.0mm ID
  • Dispense volumes from micro liters to more than 54 liters/min.
  •  
Ergonomic Design
  • Easy-grip design for use with glove-protected hands
  • Tilted head permits user to maintain a natural hand position thereby preventing hand injury
  • Momentary switch helps reduce hand fatigue by eliminating the need for constant pressure on the switch