Fisherbrand™ Exhaust Oil Recycler with Built-in Filter Element
Economical mist eliminator is recommended for use when operating the pump at a continuous vacuum pressure of 1 torr to 10 torr
Manufacturer: Fisherbrand™ 012577A
Includes: Adapters for direct connection to the pump's threaded exhaust port and gas ballast inlet port.
Description
Continuously separates air and oil from the pump exhaust, returning captured oil to the pump via the gas ballast inlet (when gas ballast valve is open). This process effectively reduces oil consumption and minimizes maintenance requirements by extending the time between oil changes.
Specifications
2-3/4 in. | |
Model M2C and M4C1 vacuum pump | |
6.25 in. | |
Exhaust Oil Recycler with Built-in Filter Element |
70 mm | |
Adapters | |
15.9 cm |