Fisherbrand™ Elite™ Multichannel Pipetters
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort
$886.45 - $1,150.35
Specifications
Products 2
Catalog Number Mfr. No. Volume (Metric) Range Number of Channels Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Range Number of Channels Price Quantity  
S98640A View Documents Fisherbrand™
4682030EMD
30 to 300μL 8 Each for $886.45
S98643A View Documents Fisherbrand™
4682070EMD
30 to 300μL 12 Each for $1,150.35
Description
Description
  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops
  • 8- and 12-channel pipetters with volume range of 1 up to 300μL
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
Specifications
Specifications
0.0
icon-pdf.png