Fisherbrand™ Electrode Connection Adapters - Mercury - Free
Adapters connect electrodes to various input jacks and are mercury free
Manufacturer:  Fisherbrand™ 13620491
 View more versions of this product
Catalog No. 13-620-491
Specifications
Specifications
US Standard/Pin Plug to BNC
Electrode
Adapts electrode with BNC plug to U.S. Standard/Pin input jack
Connection Adapter
Documents