missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Disposable Generators For Homogenizer Model 125

For processing of multiple samples per day (more than 50)

$763.30 - $763.30

Specifications

Diameter (English) Shaft 3.3 in.
Diameter (Metric) Shaft 8.4cm
Capacity (Metric) 0.3 to 10mL
For Use With (Equipment) Model 125 homogenizer
Type Disposable Generator
View More Specs

Products 1
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
08-451-679 View Documents Fisherbrand™
61030012
50/Pk. Each for $763.30
 
Description

Description

  • Needs an adaptor
  • For prevention of cross contamination

  • Dispersing plant, animal or human tissue in small amount of liquid
  • Dissolving and extraction of solids

Specifications

Specifications

3.3 in.
0.3 to 10mL
Disposable Generator
8.4cm
Model 125 homogenizer
Ideal for dispersing plant, animal or human tissue in small amount of liquid