Fisherbrand™ Digital Thermometers with Stainless-Steel Probe on Cable
Fisherbrand™ Digital thermometer with stainless-steel probe is chemically resistant to most acids, bases and solvents and may be used to measure temperature of gases, liquids, food, and other semisolids.
Manufacturer:  Fisherbrand™ CON4046
 View more versions of this product
Catalog No. 15-077-8B
Description
Description
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas. Fisher Scientific™ Digital thermometer with stainless-steel probe is chemically resistant to most acids, bases and solvents and may be used to measure temperature of gases, liquids, food, and other semisolids.
  • Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas
  • Models 15-077-16, -15, -21, -8B, -29 and 14-648-46 have alarm that can be set to alert user to temperatures that exceed setpoints
  • Some models (such as 15-077-16 and -15) have special features that print temperature, high/low memory and time of day; 15-077-29 thermometer has additional timer alarms
  • For a miniature-size thermometer, see model 15-077-32
For use in cuvets, soil, incubators, petroleum products and piercing food samples. Waterproof thermometers (models 15-077-9E, -9D and 14-648-46) are ideal for wet labs, wash-down areas and outdoor applications in the agricultural, petrochemical, food, and pharmaceutical industries.
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Specifications
−58° to 500.8°F
500.8°F
−58°F
ISO 17025, A2LA, NIST
4 x 2 x 0.5 in. (10.16 x 5.1 x 1.3cm)
0.5 in.
AAA battery
7.25 in.
304.8cm
0.1°C
5.1cm
±1°C
4 in.
−50° to 260°C
260°C
−50°C
0.12 in.
Wet labs, wash down areas and outdoor applications in the agricultural, petrochemical, food and pharmaceutical industries
1.3cm
120 in.
102mm
18.41cm
2 in.
Traceable Digital Thermometer w/Probe
Digital LCD
Documents
Accessories