Fisherbrand™ DAS-3 Data Acquisition System for Traceable™ Products with Computer Output
Powerful and easy to use computer data capture program works with Traceable Instruments with computer output—designed to work with Windows™ 98/Me/NT/2000/XP/Vista/Windows 7. Connects up to eight instruments all running simultaneously on one computer.
Manufacturer:  Fisherbrand™ CON4136
 View more versions of this product
Catalog No. 11-661-22
Includes: CD, 6 ft. cable (supplied USB, serial, and instrument connections) cable plugs into the instrument and computer.
Description
Description
Powerful and easy to use computer data capture program works with Traceable Instruments with computer output—designed to work with Windows™ 98/Me/NT/2000/XP/Vista/Windows 7. Connects up to eight instruments all running simultaneously on one computer.
  • Capture readings permanently
  • Records in intervals from 1 to 10,000 seconds
  • Straightforward plug-and-play
  • Displays minimum/maximum readings
  • Utilizes an alarm mode that permits the user to be notified visually, audibly, and by email when an alarm is triggered
  • Unlimited readings are saved to a file that can be viewed and printed as is, or may be printed in any report or spreadsheet format
  • Networking server/client capability allows the captured data to be monitored on a remote workstation
  • Readings may be streamed to cell phones or anywhere via email every minute or in any time interval
  • 24/7 notification of audible/visual alarms are sent to user's computer, remote computer, via email anywhere, and by email to cell phones
  • Program automatically installs in less than two minutes—no user programming, no user keystrokes, no user entries required: simply connect the cable to the instrument and to a USB port (or serial port) and data is captured
Specifications
Specifications
Capture readings permanently, displays minimum/maximum readings
CD, 6 ft. USB cable
Computer data acquisition system
Pressure/vacuum gauge, light meter, RTD Platinum thermometer, double thermometer, conductivity meter, humidity/thermometer, moisture meter, infrared digital thermometer, dissolved oxygen meter and hot wire anemometer/thermometer
Computer data acquisition system
Documents