Fisherbrand™ CPX Series Digital Ultrasonic Cleaning Bath
Outfit your lab with a Fisher Scientific™ classic. These tabletop cleaners offer high-tech design, solid construction and feature reliable digital controls with power tracking capabilities.
Manufacturer:  Fisherbrand™ CPX962319R
 View more versions of this product
Catalog No. 15-337-419
Description
Description
Outfit your lab with a Fisher Scientific™ classic. These tabletop cleaners offer high-tech design, solid construction and feature reliable digital controls with power tracking capabilities.
  • Sweep frequency
  • Digital controls
  • High/lower power adjustment
  • 99 minute or continuous operation, degass period up to 99 minutes
  • Constant power tracking adjusting for changes to ensure consistent performance for years of reliable use
Specifications
Specifications
1.1
40Hz
5.7L (1.5 gal.)
Cleaning
Plastic
90 min.
5.4kg
Digital
120V
29 x 15 x 15cm (11.5 x 6 x 6 in.)
Tank cover and drain
CPX3800
12 lb.
2 Years
Documents