missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Corrosion Resistant Shaft and Propellers for Overhead Stirrers

Will resist chemicals and prevent build-up.

Supplier:  Fisherbrand™ 370209037

 View more versions of this product

Catalog No. 14-512-904



Description

Description

  • Constructed with thick Virgin PTFE outer-shell
  • Rigid stainless steel inner core
  • Shaft 10mm OD x 1/4 in. Dia. core
  • Inner shaft is 1 in. longer than outer shell allowing for chucking directly on the stainless steel section of the shaft

Specifications

Specifications

0.39 in.
1 cm
Overhead Stirrer
30.48 cm
2.5 in.
6.35 cm
12 in.
Shaft with Propeller
SDS
Documents

Documents

Promotions

Promotions