Fisher Scientific™ Microscope Bundles
These two bundle options feature infinity corrected optics, Koehler illumination and the option of a 3.0 MP or 1080 HDMI camera
$3,957.29 - $5,383.06
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Camera Type Model Price Quantity  
Catalog Number Mfr. No. Camera Type Model Price Quantity  
11350131 National Optical
DC1080AX1103PL
1080 HDMI Bundle 11350131 Each for $5,383.06
11350132 National Optical
DC4AX813PL
3.0MP Bundle 11350132 Each for $3,957.29
11350133 National Optical
DC3AX813PH
3.0MP Bundle 11350133 Each for $4,872.67
Specifications
Description
  • Choose between 3.0MP camera or1080 HDMI camera
  • Video c-mount adapter (0.5X lens)
  • 3W LED Koehler illumination with intensity control
  • Built-in 160x140mm mechanical stage with extended X/Y post control
  • Inward-facing quintuple revolving nosepiece
Description & Specifications
Specifications
0.0