Please login to your online account to display your discounted pricing
Fisher Scientific™ Microscope Bundles
These two bundle options feature infinity corrected optics, Koehler illumination and the option of a 3.0 MP or 1080 HDMI camera
Manufacturer: national optical  DC3AX813PL
 View more versions of this product
Catalog No. 11350132
  • / Each
Description & Specifications
Specifications
Model Bundle 11350132
Objective Types 4X, 10X, 40X, 100X infinity corrected
Camera Type 3.0MP