missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ CMUV Series Controlled Heating Electromantles
Special “V-shaped” design accepts 60° funnels, and accomodates pear-shaped and round bottom flasks
Manufacturer: Fisherbrand™ 3601323
Description
- Large flask/funnel capacities of between 10 to 22L
- Three separate electrical circuits for complete control of heating that enable independent top, middle and bottom heating of flasks, each controlled with its own dial switch
- Stainless steel screen between flask and heating element provides spill-proof protection against electric shocks
- “Cool-to-the-touch” design
- Element temperature up to 450°C
- Indicator lamps for power and heater operation
- Replaceable insulated heater cartridge
- Deliver the benefits of the CMU Electromantle
- Easy to clean, powder coated aluminium outer casing
- Non-skid feet and three support rod clamps (not including support rod)
- Grounded earth screen and double fuses for additional safety
Certifications
CE marked
Specifications
5.8 gal., 22L | |
18.7 lb. | |
5.8 gal. |
22 L | |
8.5kg |