missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ CMUV Series Controlled Heating Electromantles

Special “V-shaped” design accepts 60° funnels, and accomodates pear-shaped and round bottom flasks

Manufacturer:  Fisherbrand™ 3601319

 View more versions of this product

Catalog No. 11-747-313



Description

Description

  • Large flask/funnel capacities of between 10 to 22L
  • Three separate electrical circuits for complete control of heating that enable independent top, middle and bottom heating of flasks, each controlled with its own dial switch
  • Stainless steel screen between flask and heating element provides spill-proof protection against electric shocks
  • “Cool-to-the-touch” design
  • Element temperature up to 450°C
  • Indicator lamps for power and heater operation
  • Replaceable insulated heater cartridge

  • Deliver the benefits of the CMU Electromantle
  • Easy to clean, powder coated aluminium outer casing
  • Non-skid feet and three support rod clamps (not including support rod)
  • Grounded earth screen and double fuses for additional safety

Certifications

CE marked

Specifications

Specifications

3.1 gal., 12L
13.2 lb.
3.1 gal.
12 L
6kg
SDS
Documents
Product Certifications
Promotions

Promotions