Fisherbrand™ CMUT Series Electromantles
Adjustable volume able to accommodate round bottom flasks between 250 to 1000mL capacity
Supplier:  Fisherbrand™ 3601315DUP
Catalog No. 36-101-2692
Description
Description
  • Element temperature up to 450°C
  • Cool-to-the-touch design
  • Built in energy regulator
  • Power and heating operation indicated via indicator lamps
  • Replaceable heater cartridge
  • Grounded stainless-steel earth screen and double fuses for added safety
  • Used in wet chemistry to heat liquids in round bottom flasks.
  • Non-skid feet and support clamps add safety
Certifications
CE marked
Warranty
One year (Part and Labour)