missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Scientific™ CMUA Stirring Mantles

Available in sizes 50 to 5000mL

Manufacturer:  Fisher Scientific™ 3601305

 View more versions of this product

Catalog No. 11-474-82


 

Specifications Description & Specifications

Specifications

115V
CE, CSA
2000rpm
450°C
1.5kg
500mL
500mL
115V
3.3 lb.
16.9 oz.
8.66 in.
842°F
280w