Please login to your online account to display your discounted pricing
Fisherbrand™ Basic Cleanroom/ESD Chair, Desk Height, Chrome, Vinyl Seat
Choose Fisherbrand Cleanroom/ESD chairs to provide proper grounding and dissipative static control in your laboratory.
$1,068.29 - $1,068.29
Specifications
Adjustable (Height) 18 to 23 in.
Frame Type Chrome
Seat Material Vinyl Upholstered Foam
Footrest None
Foot Type Casters
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
14-359-918
e com
91267PBKCCR100CABES
Black Each for $1,068.29
14-359-970
e com
91267PBCCR100CABESD
Blue Each for $1,068.29
Description & Specifications
Specifications
Adjustable (Height) 18 to 23 in.
Frame Type Chrome
Seat Material Vinyl Upholstered Foam
Footrest None
Foot Type Casters
Adjustable Standard Lift
  • Cleanroom/ESD chairs have been tested to Fed Std.209E, ISO 14644-1 (Class10/ISO Class 4), and ANSI/ESD-STM-12.1-2006 standards and feature proper grounding, conductive, and dissipative static control materials
  • Adjusts from 18 to 23 inches
15-year warranty, lifetime on the pneumatic cylinder.