Please login to your online account to display your discounted pricing

Fisher Scientific™ Classroom Periodic Charts


Two-color charts conform to the recommendations of the IUPAC.

Manufacturer: fisher scientific™  S45520

 View more versions of this product

Catalog No. S45520

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Product Type Periodic Table Chart: Notebook size
Certifications/Compliance IUPAC regulations
Classification Science
For Use With (Application) Long form chart shows elements, their atomic weights, numbers and the number of electrons in each shell
Height (English) 11 in.
Width (English) 8.5 in.