Fisherbrand™ Baume Hydrometers
Baume “heavy” scale for working with liquids heavier than water.
Supplier: Fisherbrand™ 11580C
Description
Baume “heavy” scale for working with liquids heavier than water.
- Permanently marked with individual serial numbers
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Specifications
Baume Hydrometer | |
Without 3-point certification | |
Soda Lime Glass | |
Baume 'Heavy' Scale For Liquids Heavier than Water |
1° | |
285 mm | |
0 to 70° |