Basix™ Syringe Filters, PVDF, Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisherbrand™ Basix™ Syringe Filters, PVDF, Sterile offers high sample throughput.
$105.33 - $105.33
Specifications
Membrane PVDF
Sterility Sterile
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Pore Size Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Pore Size Price Quantity  
13100104
View Documents
Fisherbrand
13100104
25mm 0.2μm
Pack of 50 Each for $105.33
Case of 10 Pack for $1,053.30
13100105
View Documents
Fisherbrand
13100105
25mm 0.45μm
Pack of 50 Each for $105.33
Case of 10 Pack for $1,053.30
Description
Description
  • Color-coded for easy identification
  • Available in two different diameter to suit the applications
  • Individually wrapped
Specifications
Specifications
PVDF
Sterile
SDS