Basix™ Syringe Filters, PTFE, Non-Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisherbrand™ Basix™ Syringe Filters, PTFE, Non-Sterile offers high sample throughput.
 View more versions of this product
Catalog No. 13100115
Description
Description
  • Color-coded for easy identification
  • Available in two different diameter and pore size to suit the applications
Specifications
Specifications
Hydrophobic PTFE
Non-sterile
25 mm
0.2 μm
Polypropylene