missing translation for 'onlineSavingsMsg'
Learn More

Basix™ Syringe Filters, PES, Sterile

Catalog No. 13100107
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
13100107 0.45 μm
13100106 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13100107
Edge

Attach to the end of syringes and filter contents as it is being forced out. Fisherbrand™ Basix™ Syringe Filters, PES, Sterile offers high sample throughput.

  • Color-coded for easy identification
  • Available in two different diameter to suit the applications
  • Individually wrapped 

Specifications

Material (Membrane) PES
Pore Size 0.45 μm
Sterility Sterile
Housing Material Acrylic
Diameter (Metric) 25 mm