Fisher Scientific™ AX800 Series Binocular Compound Microscopes
Ergonomically designed, professional frame with low-position coaxial focusing allows one-handed operation
Manufacturer:  National Optical AX813PL
 View more versions of this product
Catalog No. 11350112
Specifications Description & Specifications
Specifications
IEC Power Cord, Built-in Cord Hanger, Dustcover, Allen Key
Z-Axis Movement
Infinity Corrected 4X, 10X, 40X, 100X
University and laboratory use
Interpupillary Distance 55-75mm
Reversed quintuple revolving nosepiece
IEC 110/220V (UL / CSA)
11350112
Focusable Abbe Condenser N.A. 1.25 with iris diaphragm
N-WF 10X/20mm with diopter adjustment, ±5 diopter (Rubber eyeshields included)
Trinocular
3W Koehler LED illumination with intensity control
7kg
5 year limited warranty, excluding bulbs, cords, and fuses
Accessories