Fisher Scientific™ AP Lab 1: Diffusion and Osmosis
Investigate diffusion and osmosis using a model membrane system.
Manufacturer:  Fisher Science Education AZ-20-2603
 View more versions of this product
Catalog No. S66052
Specifications Description & Specifications
Specifications
8 lab stations
Diffusion, osmosis, water potential, calculation of water potential from experimental data, onion cell plasmolysis
Investigate diffusion and osmosis using a model membrane system, observe how the process of diffusion allows the cell to absorb vital nutrients and eliminate wastes using a semi-permeable membrane model that simulates a living cell
Ap™ Biology Lab 1: Diffusion And Osmosis