Fisher Scientific™ AP Lab 1: Diffusion and Osmosis
Investigate diffusion and osmosis using a model membrane system.
$189.00 - $189.00
Specifications
For Use With (Application) Investigate diffusion and osmosis using a model membrane system, observe how the process of diffusion allows the cell to absorb vital nutrients and eliminate wastes using a semi-permeable membrane model that simulates a living cell
View More Specs
Includes: Activities include:
1a. Diffusion
1b. Osmosis
1c. Water potential
1d. Calculation of water potential from experimental data
1e. Onion cell plasmolysis
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
S66052 Fisher Science Education
AZ-20-2603
Diffusion, osmosis, water potential, calculation of water potential from experimental data, onion cell plasmolysis Ap™ Biology Lab 1: Diffusion And Osmosis Each for $189.00
Specifications
Description
•    Comprehensive background section covers mechanisms of diffusion and osmosis, selectively-permeable membranes, concept of water potential, and concept of molarity, as well as its relationship to osmotic concentration
•    Students will observe how the process of diffusion allows the cell to absorb vital nutrients and eliminate wastes using a semi-permeable membrane model that simulates a living cell
•    The class will also study osmosis by measuring the water potential of a solution
•    Curriculum Pack Includes the Eight Lab Station Investigation and the individual CD-ROM.
Description & Specifications
Specifications
Investigate diffusion and osmosis using a model membrane system, observe how the process of diffusion allows the cell to absorb vital nutrients and eliminate wastes using a semi-permeable membrane model that simulates a living cell