Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
Manufacturer:  Fisherbrand™ 370209014
 View more versions of this product
Catalog No. 14-500-216
Description
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
  • Can be used with flammable chemicals for explosion proof mixing
Specifications
Specifications
0.5 in.
10 lb.
1.27cm
1.5
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
1690.7 oz.
50L
3000rpm
4.6kg
Safely mixing all types of solvents, lacquers and volatile chemicals
40 to 100psi
633
50000mL
30000