Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
Manufacturer:  Fisherbrand™ 370209016
 View more versions of this product
Catalog No. 14-500-218
Description
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
  • Can be used with flammable chemicals for explosion proof mixing
Specifications
Specifications
0.37 in.
6 lb.
0.95cm
0.3
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
676.28 oz.
20L
1200rpm
2.7kg
Safely mixing all types of solvents, lacquers and volatile chemicals
30 to 80psi
203
20000mL
10000