Fisherbrand™ accuTupH™ Rugged Bulb pH Indicating Electrode - Mercury - Free
Can be optimal for lab use and precise Q.C. measurements.
Manufacturer:  Fisherbrand™ 13620186
 View more versions of this product
Catalog No. 13-620-186
Description
Description
  • 0 to 14pH range
  • Available with BNC or U.S. Standard connectors
Specifications
Specifications
U.S. Standard
10mm
32° to 212°F
5.62 in.
0° to 100°C
0.37 in.
0 to 14
0° to 100°C
143mm
Documents