Please login to your online account to display your discounted pricing
Fisher Scientific™ accumet™ XL Benchtop Meters
High-quality, user-friendly and cost-effective
$3,123.67 - $8,524.26
Specifications
Accuracy Multiple
Accuracy (mV) ±0.2mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.3°C
Certifications/Compliance NIST, ISO, FDA, USDA, EPA
View More Specs
Products
Catalog Number Mfr. No. Includes Inputs Model Price Quantity    
13-636-XL150A
  Accessories
fisher scientific™
13636XL150A
Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply BNC, ATC XL150 pH Meter Each for $3,123.67
13-636-XL150
  Accessories
fisher scientific™
13636XL150
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply BNC, ATC XL150 pH Kit Each for $3,449.81
13-636-XL250
  Accessories
fisher scientific™
13636XL250
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL250 Dual pH/ISE Kit Each for $4,216.42
13-636-XL200A
  Accessories
fisher scientific™
13636XL200A
Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply BNC, ATC XL200 pH/Conductivity Meter Each for $4,000.42
13-636-XL200
  Accessories
fisher scientific™
13636XL200
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp. probe (13-620-100), electrode arm, RS-232 and USB cables, and 110/220V power supply BNC, ATC XL200 pH/Conductivity Kit Each for $4,561.61
13-636-XL250F
  Accessories
fisher scientific™
13636XL250F
Meter, 13-620-629 fluoride electrode, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL250 Dual pH/ISE Fluoride Kit Each for $4,735.25
13-636-XL250N
  Accessories
fisher scientific™
13636XL250N
Meter, 13-620-509 ammonia electrode, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL250 Dual pH/ISE Ammonia Kit Each for $5,599.31
13-636-XL250A
  Accessories
fisher scientific™
13636XL250A
Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL250 Dual pH/ISE Meter Each for $3,871.22
13-636-XL500
  Accessories
fisher scientific™
13636XL500
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp. probe (13-620-100), electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL500 Dual pH/ISE/Conductivity Kit Each for $5,664.97
13-636-XL500A
  Accessories
fisher scientific™
13636XL500A
Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL500 Dual pH/ISE/Conductivity Meter Each for $5,080.44
13-636-XL600A
  Accessories
fisher scientific™
13636XL600A
Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL600 Dual pH/ISE/Conductivity/DO Meter Each for $6,032.86
13-636-XL600D
  Accessories
fisher scientific™
13636XL600D
Meter, self-stirring BOD probe (13-620-SSP), TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp probe (13-620-100), electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL600 Dual pH/ISE/Conductivity/DO Deluxe Kit Each for $7,976.50
13-636-XL600
  Accessories
fisher scientific™
13636XL600
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp. probe (13-620-100), electrode arm, RS-232 and USB cables, and 110/220V power supply 2 BNC, 2 ATC XL600 Dual pH/ISE/Conductivity/DO Kit Each for $6,486.31
XL200ACERT
  Accessories
fisher scientific™
59331-09
Meter, electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate BNC, ATC XL200 pH/Conductivity Meter w/ NIST-Traceable Certificate Each for $5,326.15
XL250ACERT
  Accessories
fisher scientific™
59331-11
Meter, electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate 2 BNC, 2 ATC XL250 Dual pH/ISE Meter w/ NIST-Traceable Certificate Each for $5,137.26
XL250CERT
  Accessories
fisher scientific™
59331-12
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate 2 BNC, 2 ATC XL250 Dual pH/ISE Kit w/ NIST-Traceable Certificate Each for $5,577.94
XL200CERT
  Accessories
fisher scientific™
59331-10
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp probe (13-620-100), electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate BNC, ATC XL200 pH/Conductivity Kit w/ NIST-Traceable Certificate Each for $5,993.44
XL150CERT
  Accessories
fisher scientific™
59331-08
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate BNC, ATC XL150 pH Kit w/ NIST-Traceable Certificate Each for $4,646.15
XL150ACERT
  Accessories
fisher scientific™
59331-07
Meter, electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate BNC, ATC XL150 pH Meter w/ NIST-Traceable Certificate Each for $4,255.88
XL500ACERT
  Accessories
fisher scientific™
59331-13
Meter, electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate 2 BNC, 2 ATC XL500 Dual pH/ISE/Conductivity Meter w/ NIST-Traceable Certificate Each for $6,585.24
XL500CERT
  Accessories
fisher scientific™
59331-14
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp probe (13-620-100), electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate 2 BNC, 2 ATC XL500 Dual pH/ISE/Conductivity Kit w/ NIST-Traceable Certificate Each for $7,277.76
XL600CERT
  Accessories
fisher scientific™
59331-17
Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp probe (13-620-100), electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate 2 BNC, 2 ATC XL600 Dual pH/ISE/Conductivity/DO Kit w/ NIST-Traceable Certificate Each for $8,524.26
XL600ACERT
  Accessories
fisher scientific™
59331-16
Meter, electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate 2 BNC, 2 ATC XL600 Dual pH/ISE/Conductivity/DO Meter w/ NIST-Traceable Certificate Each for $7,970.24
XL600DCERT
  Accessories
fisher scientific™
59331-18
Meter, self-stirring BOD probe (13-620-SSP), TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp probe (13-620-100), electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate 2 BNC, 2 ATC XL600 Dual pH/ISE/Conductivity/DO Deluxe Kit w/ NIST-Traceable Certificate CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Accuracy Multiple
Accuracy (mV) ±0.2mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.3°C
Certifications/Compliance NIST, ISO, FDA, USDA, EPA
Data Management Details 3 to 86400 sec.
Detection Range Multiple
Calibration Points (pH) Up to 6 pre-set or 5 custom
Data Points 2000
Range (pH) -2 to 20
Resolution (pH) 0.1/0.01/0.001
Display Type LCD
Height (English) 3.89 in.
Height (Metric) 9.9cm
Length (English) 9.68 in.
Length (Metric) 24.6cm
Outputs RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
Range (mV) ±2000mV
Resolution Multiple
Resolution (mV) 0.1mV
Resolution (Temperature) 0.1°C
Voltage 100/240V
Warranty 3 Years
Weight (English) 2.5 lb.
Weight (Metric) 1.13kg
Width (Metric) 17.3cm
Fisher Scientific accumet XL benchtop meters bring advanced features with simplicity to the laboratory. The color-touchscreen is stunningly vivid and is sure to make a great first impression that doesn't stop. Newly added features include two stirring probe ports, a 3-position electrode holder, upgradeable software, and USB & RS232 connectivity. Prices have been lowered, so these advanced XL meters have never been more affordable.
  • High–resolution color display provides crystal clear data from wide viewing angles and at a distance
  • Improved touch–screen with glass/glass sensitivity provides unmatched ease of operation and setup
  • Optional stirring probe accessory provides mixing at 5 speeds without a stir plate; connect one or two stirring probes
  • RJ45 Ethernet/Internet port—connect to Local Area Network (LAN) or Internet, print data to a network USB host connectivity for peripherals such as keyboard, mouse, or other USB device
  • Compact RS-232 printer (13-637-690) available or use with widely available USB printers such as Dell™ 1130N
  • Every instrument includes one electrode arm for use in three different positions; Every instrument can accept another electrode arm to perform measurements, and from either or both sides
  • Optional calibration reminder & high/low alarm
  • Allows up to 10 password protected ID's; each includes memory to store 2000 data points
  • Base ID's on individual users, applications, or projects to keep data separate, and retain unique calibration and instrument settings
  • Setup screens allow for simple but extensive customization
  • Calibrate with custom pH buffer values or automatically from one of six preset buffer groups including pure water buffer group
  • Iincreased 2000 point memory for up to 10 user ID's—add data manually or at specified intervals
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Preferred language setting displayed on the instrument
  • 100-240 V power supply includes multi-plugs for use around the globe
  • Wall–mounting back panel creates additional bench space
  • Date and Time to meet GLP
  • User prompts and context–specific “help” screens guide you through operation
For XL200, XL500, XL600 (Conductivity)
  • All XL200, XL500, XL600 kits now include 13-620-100 cell
  • For XL600 (DO)
    • Display up to four measurements (2 × BNC, Conductivity, & DO) simultaneously
    • DO mode calculates five-day BOD testing, OUR, and SOUR
    • Salinity correction and automatic barometric pressure compensation with built–in barometer enhance accuracy
    For XL250, XL500, XL600 (Ion Selective)
    • Dual–BNC channels; use them for pH or mV or ion selective measurement
    • Reads ISE in ppm, %, mg/mL, and mole/L
    • Direct and indirect ISE measurements, including known addition and subtraction methods, and analyte
    • Two new XL250 kits available that include an ammonia or fluoride ion selective electrode
    NIST-Traceable Calibration Models (-CERT)
    • Helps meet ISO, FDA, USDA and EPA quality assurance guidelines
    • Includes measurement of meter accuracy at five test points across pH range
    • Includes measurement of meter accuracy at five test points across corresponding mV range
    • Testing performed by accredited laboratory; includes NIST-Traceable certificate
    Select models available with Certificate of NIST Traceability
    Meters: Three years parts and labor; Electrodes (included with meter-kits): One year parts and labor