missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ accumet™ Polymer Membrane Combination ISEs, Mercury-Free

Fast, precise measurements of calcium, nitrate and potassium ion in aqueous solutions over a wide range of concentrations

Manufacturer:  Fisherbrand™ 13620532

 View more versions of this product

Catalog No. 13-620-532



Description

Description

Sensor sealed in epoxy body

  • Sensing modules contain liquid internal filling solution in contact with gelled organophilic membrane
  • Membrane is impregnated with a selective ion exchanger for ion of interest
  • Sensors are sealed into durable epoxy body with built-in reference half-cell
  • Permit fast, precise measurements in aqueous solutions over a wide range of concentrations
  • Epoxy body provides chemical resistance
Features
  • Tough, chemical-resistant epoxy body
  • Membrane assembly sealed to body
  • Continuous electrical shielding and insulation of internal elements, cable and connectors
  • Extremely stable, reproducible readings with a minimum of electrical noise
  • Lower detection limits than conventional ISEs
  • Rapid response time that lowers cost per test
  • Individually tested, serialized to meet GLP requirements and backed by the Fisher Scientific Technical Applications staff
Dimensions
  • Diameter: body, 0.5 in. (12.7mm); cap, 0.625 in. (16mm)
  • Length: body, 4 in. (10.2cm); cap, 1 in. (2.5cm); overall, 5 in. (12.7cm)

Calcium Electrode

  • Finds application in the testing of wine, beer, dairy products, foods, plants, pharmaceuticals, fertilizers, explosives, clay and soil
  • Used as an endpoint detector in EDTA calcium titrations
  • Filling solution is 4M KCl (with AgCl)
Nitrate Electrode
  • Used in pollution testing, bioresearch, pharmaceutical production
  • Determines nitrate in soils, plants, sewage, fertilizers, explosives, meats, pickling baths
  • Filling solution is 0.1M (NH4)2SO4/0.01M KCl (with Ag+)
Potassium Electrode
  • Aids in analysis of serum, urine, saliva, other bodily fluids
  • Used in testing soils, sewage, fertilizers, foods and beverages
  • Filling solution is 5M NaCl

Specifications

Specifications

Used in testing soils, sewage, fertilizers, foods, beverages, analysis of serum, urine, saliva and other bodily fluids
0.5 in.
15.8mm
5 in.
Epoxy Body
32° to 88°F
2.5 to 11
10 to 20megohm
Potassium
BNC
12.7mm
4mol/L KCl with AgCl
127mm
0.04 to 39000ppm
0° to 40°C
Cs+, NH4, Tl+, H+, Ag+, Tris+, Li+, Na+
∽1 min. or less
0 to 40°C
Documents
Accessories