missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ accumet™ Polymer Membrane Combination ISEs, Mercury-Free
Fast, precise measurements in aqueous solutions over a wide range of concentrations.
$2,010.06 - $2,010.06
Specifications
Diameter (English) Body | 0.5 in. |
---|---|
Diameter (Metric) Body | 12.7mm |
Diameter (Metric) Cap | 15.8mm |
Connector Types | BNC |
Material | Epoxy Body |
Catalog Number | Mfr. No. | Filling Solution Type | pH Range | Resistance | Electrode | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Filling Solution Type | pH Range | Resistance | Electrode | Price | Quantity | ||
13-620-534
|
Fisherbrand™
13620534 |
0.1M (NH4)2SO4/0.01mol/L KCl with Ag+ | 2 to 12 | 100megohm | Nitrate |
Each for $2,010.06
|
|
Description
Fast, precise measurements in aqueous solutions over a wide range of concentrations.
Sensor sealed in epoxy body
- Sensing modules contain liquid internal filling solution in contact with gelled organophilic membrane
- Membrane is impregnated with a selective ion exchanger for ion of interest
- Sensors are sealed into durable epoxy body with built-in reference half-cell
- Permit fast, precise measurements in aqueous solutions over a wide range of concentrations
- Epoxy body provides chemical resistance
- Tough, chemical-resistant epoxy body
- Membrane assembly sealed to body
- Continuous electrical shielding and insulation of internal elements, cable and connectors
- Extremely stable, reproducible readings with a minimum of electrical noise
- Lower detection limits than conventional ISEs
- Rapid response time that lowers cost per test
- Individually tested, serialized to meet GLP requirements and backed by the Fisher Scientific Technical Applications staff
- Diameter: body, 0.5 in. (12.7mm); cap, 0.625 in. (16mm)
- Length: body, 4 in. (10.2cm); cap, 1 in. (2.5cm); overall, 5 in. (12.7cm)
Calcium Electrode
- Finds application in the testing of wine, beer, dairy products, foods, plants, pharmaceuticals, fertilizers, explosives, clay and soil
- Used as an endpoint detector in EDTA calcium titrations
- Filling solution is 4M KCl (with AgCl)
- Used in pollution testing, bioresearch, pharmaceutical production
- Determines nitrate in soils, plants, sewage, fertilizers, explosives, meats, pickling baths
- Filling solution is 0.1M (NH4)2SO4/0.01M KCl (with Ag+)
- Aids in analysis of serum, urine, saliva, other bodily fluids
- Used in testing soils, sewage, fertilizers, foods and beverages
- Filling solution is 5M NaCl
Specifications
0.5 in. | |
15.8mm | |
Epoxy Body | |
127mm | |
32° to 88°F | |
0 to 40°C |
12.7mm | |
BNC | |
5 in. | |
∽1 min. or less | |
0° to 40°C | |
Cl2-, NO-, Br-, CN-, CIO-3, I-, CIO4- |