Fisherbrand™ accumet™ pH Indicating Glass Body Universal Glass Membrane Models- Mercury-Free
Universal glass membrane and glass body provides measurements from zero to 14pH
Manufacturer:  Fisherbrand™ 136204
 View more versions of this product
Catalog No. 13-620-4
Description
Description
  • Universal glass membrane
  • Zero to 14pH range
Specifications
Specifications
U.S. Standard
0 to 14
-5° to 110°C
60 in.
Glass
23° to 230°F
Titralyzer II
Suggested Reference Electrodes: 13-620-64
Documents