missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ accumet™ Micro and Semimicro Size Glass Combination Electrodes - Mercury - Free

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Manufacturer:  Fisherbrand™ 13620292

 View more versions of this product

Catalog No. 13-620-292



Description

Description

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Zero to 14pH range
Specifications

Specifications

-5° to +110°C
0 to 14
Glass
Saturated KCl with AgCl. KCl
6mm
Ag/AgCl
23° to 230°F
0.23 in.
pH/ORP combination titration electrode, Extra-Long Ag/AgCl
1 Year
-5° to 110°C
Single
150mm
BNC
7 in.
Small samples
Micro
Documents
Accessories