missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ accumet™ Micro and Semimicro Size Glass Combination Electrodes - Mercury - Free

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Manufacturer:  Fisherbrand™ 1362092

 View more versions of this product

Catalog No. 13-620-92



Description

Description

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Zero to 14pH range
Specifications

Specifications

-5° to +110°C
0 to 14
Glass
Saturated KCl with AgCl. KCl
U.S. Standard
23° to 230°F
Micro
1 Year
-5° to 110°C
Single
100mm
Ag/AgCl
Small samples
pH/ORP combination titration electrode, Standard Ag/AgCl
Documents