missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free

For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical

Manufacturer:  Fisherbrand™ 1362091

 View more versions of this product

Catalog No. 13-620-91



Includes: Removable bulb protector

Description

Description

For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical

  • 0 to 14pH range
  • Porous ceramic junction
  • Twist-open/twist-close cap for easy filling
  • Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
  • Immersion depth, ∽4 in. (10cm)
  • 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in.L (152.4cm) lead

Choice of Ag/AgCl Single- or Double-Junction internal reference elements

  • Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
  • Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers

  • For aqueous and nonaqueous samples
  • Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers

Specifications

Specifications

Blue
1 Year
23° to 212°C
Single Ceramic
101.6mm
U.S. Standard/pin
Ag/AgCl
60 in.
For routine pH and titration measurements with small samples and small vessels, where separate half cell electrodes may be impractical
pH combination electrode
<10 sec.
-5 to +100°C
0 to 14
Glass
SP135-500
152.4cm
pH meter
4 in.
-5° to +100°F
Removable bulb protector
Twist open/Twist close
Documents