Please login to your online account to display your discounted pricing

Fisher Scientific™ accumet™ Glass Body Calomel Reference Electrodes

$435.76 - $435.76

Specifications

Diameter (English) Body 0.37 in.
Diameter (Metric) Body 1cm
Connector Types Pin
Material Glass
Filling Solution Type Mercury
View More Specs

 Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Products
Catalog Number Mfr. No. Applications Diameter (Metric) Cap Length (English) Price Quantity    

13-620-57

 
fisher scientific™
1362057
Biological, Acid, Nonaqueous Solutions 1.6cm 5.75 in. Each for $435.76
Description & Specifications

Specifications

Diameter (English) Body 0.37 in.
Diameter (Metric) Body 1cm
Connector Types Pin
Material Glass
Filling Solution Type Mercury
Internal Reference Hg/Hg2Cl2 (Calomel)
Length (English) Cable 30 in.
Length (Metric) 14.6cm
Length (Metric) Cable 76.2cm
pH Range 0 to 14
Reference Electrolyte Calomel
Temperature (English) Operating 23° to 176°F
Temperature (Metric) Operating -5° to +80°C

  • Standard-size glass body
  • Contains mercury
  • -5° to +80°C
  • 0 to 14pH range

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.