Please login to your online account to display your discounted pricing
Fisher Scientific™ accumet™ Epoxy Body Mercury-Free Reference Electrode
A rugged epoxy body makes this a great choice for demanding environments
$369.56 - $394.88
Products
Catalog Number Mfr. No. Price Quantity    
13-620-858
  Accessories
fisher scientific™
13620858
Each for $369.56
13-620-859
fisher scientific™
13620859
Each for $394.88
Description & Specifications
  • Rugged epoxy body for in-plant monitoring, field measurements, instructional labs, classrooms
  • Stable with ion-selective electrodes
  • Tough enough to hand hold, use directly in a vessel, stir solutions
  • No need for auxiliary stirrer
Use with indicating cell.