Fisherbrand™ accumet™ Epoxy Body Mercury-Free Reference Electrode
A rugged epoxy body makes this a great choice for demanding environments
Manufacturer:  Fisherbrand™ 13620859
 View more versions of this product
Catalog No. 13-620-859
Description
Description
  • Rugged epoxy body for in-plant monitoring, field measurements, instructional labs, classrooms
  • Stable with ion-selective electrodes
  • Tough enough to hand hold, use directly in a vessel, stir solutions
  • No need for auxiliary stirrer
Use with indicating cell.
Specifications
Specifications
Field, lab
Gel
30 in.
Epoxy
Pin
Porous polymer
76.2cm
Documents