Please login to your online account to display your discounted pricing
Fisher Scientific™ accumet™ Dot-Matrix Printer
Impact dot-matrix printer for accumet AR Series Benchtop Meters
$47.10 - $47.10
Specifications
For Use With Accumet™ AR50, AR25, AR20, AR15 benchtop meter
View More Specs
Includes:  13-637-667 Serial AR Printer Cable
Products
Catalog Number Mfr. No. Type Price Quantity    
13-637-668
fisher scientific™
3597668
Printer Ribbon Each for $47.10
Description & Specifications
Specifications
For Use With Accumet™ AR50, AR25, AR20, AR15 benchtop meter
  • Permanently records results on 2.25 in. (5.7cm) paper with an ink ribbon
  • Prints up to 1.625 lines per second, 28.75 characters per line
  • Operates with large (12,500 lines) or small (3000 lines) roll paper
  • Print speed 38 lines per minute for 24-column
  • Printer communicates via standard RS-232-C interface (also includes RS485 port)
  • Baud rate is 19,200bps
Electrical Requirements: 120V 60Hz 20w (input to outlet); 9V 1.5A (output to printer). Stepdown converter included. Operating temperature, 5° to 40°C (41° to 104°F). Dimensions, 4.125L × 4.5W × 2 in. H (10.4 × 11.4 × 5cm).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.