Please login to your online account to display your discounted pricing

Fisher Scientific™ accumet™ Conductivity Cells for AP65 Portable Conductivity Meter - Mercury - Free


Manufacturer: fisher scientific™  13620167

 View more versions of this product

Catalog No. 13-620-167

  • / Each
 

Description & Specifications

Specifications

Product Type Conductivity Cell
Cell Constant 0.1cm-1
For Use With Accumet AP65 portable conductivity meter
Material Epoxy
Measurement Ranges 0.5 to 200μS
Storage Method Immersed in deionised water
Probe Two cell/ATC probe

  • Epoxy body
  • Two-cell/ATC conductivity probe
  • Cell Constant:  0.1cm-1
  • Range: 0.5 to 200μS/cm