Please login to your online account to display your discounted pricing
Fisher Scientific™ accumet™ Conductivity Cells for AP65 Portable Conductivity Meter - Mercury - Free
$771.48 - $900.82
Products
Catalog Number Mfr. No. Price Quantity    
13-620-168
fisher scientific™
13620168
Each for $771.48
13-620-167
fisher scientific™
13620167
Each for $900.82
Description & Specifications
  • Immersion-type cells feature the same compact size of a typical pH electrode, reducing the necessary sample size for analysis
  • Epoxy body construction stands up to rugged handling and usage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.