missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Scientific™ accumet™ AB150 pH Benchtop Meters

Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisher Scientific accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.

Manufacturer:  Fisher Scientific™ 5933100

 View more versions of this product

Catalog No. AB150ACERT


 

Includes: accumet AB150 pH benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST traceability certificate

Specifications

Description

  • Graphic backlit display with date and time, stability indicator and alarms
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • One point relative mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
NIST-Traceable Calibration Models (-CERT)
  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory; includes NIST-Traceable certificate
Specifications
  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial

Warranty

3 years on meter

Description & Specifications

Specifications

3 Years
-2 to 20
±0.002
0.1mV
Up to 6 pre-set or 5 custom
±0.3°C
Manual, timed (selectable every 3 to 3600 sec.), printer or CSV format
BNC, ATC
With NIST-traceable pH Certificate
AB150 pH Meter Only
Meter, electrode arm, RS-232 and USB cables, 100/240V power supply, and NIST traceability certificate
0.1/0.01/0.001
±2000mV
±0.2mV
0.1°C
LCD
500
RS232 (phono plug), mini-B USB, stirrer
50/60Hz
100/240V