Please login to your online account to display your discounted pricing
Fisher Scientific™ accumet™ AB Benchtop Meters
Available with ISE and multi-parameter pH/Conductivity capabilities
$1,648.10 - $4,495.06
Specifications
Accuracy (mV) ±0.2mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.3°C
Range (pH) -2 to 20
Resolution (pH) 0.1/0.01/0.001
View More Specs
Products
Catalog Number Mfr. No. Certifications/Compliance Includes Model Price Quantity    
13-636-AB150A
  Accessories
fisher scientific™
13636AB150A
Each for $1,925.08
13-636-AB150
  Accessories
fisher scientific™
13636AB150
Each for $2,162.11
13-636-AB200
  Accessories
fisher scientific™
13636AB200
Each for $3,338.56
13-636-AB150B
  Accessories
fisher scientific™
13636AB150B
Each for $1,648.10
13-636-AB250
fisher scientific™
13636AB250
Each for $2,162.92
13-636-AB250A
fisher scientific™
13636AB250A
Each for $1,925.08
AB150ACERT
  Accessories
fisher scientific™
59331-00
With NIST-traceable pH Certificate Meter, electrode arm, RS-232 and USB cables, 100/240V power supply, and NIST traceability certificate AB150 pH Meter Only Each for $2,694.56
AB150BCERT
  Accessories
fisher scientific™
59331-02
With NIST-traceable pH Certificate Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, 110/220V power supply, and NIST traceability certificate AB150 pH BioBasic Kit Each for $3,338.68
AB200ACERT
fisher scientific™
59331-05
With NIST-traceable pH Certificate Meter, electrode arm, RS-232 and USB cables, 100/240V power supply, and NIST traceability certificate AB200 pH/Conductivity Meter Only Each for $3,975.26
AB150CERT
  Accessories
fisher scientific™
59331-01
With NIST-traceable pH Certificate Meter, TRIS compatible pH/ATC electrode (13-620-631), electrode arm, RS-232 and USB cables, 100/240V power supply, and NIST traceability certificate AB150 pH Kit Each for $3,065.88
AB200CERT
fisher scientific™
59331-06
With NIST-traceable pH Certificate Meter, TRIS compatible pH/ATC electrode (13-620-631), Conductivity/Temp probe (13-620-100), electrode arm, RS-232 and USB cables, 100/240V power supply, and NIST traceability certificate AB200 pH/Conductivity Kit Each for $4,495.06
AB250ACERT
fisher scientific™
59331-03
With NIST-traceable pH Certificate Meter, electrode arm, RS-232 and USB cables, 100/240V power supply, and NIST traceability certificate AB200 pH/Conductivity Meter Only Each for $3,442.59
AB250CERT
fisher scientific™
59331-04
With NIST-traceable pH Certificate Meter, TRIS compatible pH/ATC electrode (13-620-631), electrode arm, RS-232 and USB cables, 100/240 V power supply, and NIST traceability certificate AB250 pH/ISE Kit Each for $3,728.41
Description & Specifications
Specifications
Accuracy (mV) ±0.2mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.3°C
Range (pH) -2 to 20
Resolution (pH) 0.1/0.01/0.001
Display Type LCD
Range (mV) ±2000mV
Resolution (mV) 0.1mV
Voltage 100/240V
Warranty 3 Years
Intuitive, simple operation and high accuracy in a compact, affordable meter.
  • Optional stirring probe accessory (Cat. No. 13-620-BSP) with five speed settings
  • Compact size conserves valuable bench space
  • Large display shows measurement and temperature at all times
  • Automatic Temperature Compensation (ATC) in °C or °F
  • Graphic display with adjustable backlight
  • Date and Time to meet GLP
  • Password protection
  • Optional calibration reminder and high/low alarm
  • 500-point memory — log manually or at intervals
  • Easy-to-understand prompts
  • Easily view calibration data
  • 100 to 240V power supply includes multi-plugs for use around the globe
  • Select preferred language as displayed on instrument — choose from English, German, Chinese, French, Italian and Spanish
For AB200 only
  • Measures pH and Conductivity/TDS/Resistivity/Salinity
  • Temperature is measured in two channels
  • High accuracy thermistor built in to the conductivity probe, or separately along with the pH reading
For AB150, 250 only
  • 0.001pH resolution
  • Calibrate with user-selected custom pH buffer values
For AB250 only
  • Measures pH and Ion Selective
  • NIST-Traceable Calibration Models (-CERT)
    • Helps meet ISO, FDA, USDA and EPA quality assurance guidelines
    • Includes measurement of meter accuracy at five test points across pH range
    • Includes measurement of meter accuracy at five test points across corresponding mV range
    • Testing performed by accredited laboratory; includes NIST-Traceable certificate
    Select models available with Certificate of NIST Traceability
    Meters: Three years parts and labor; Electrodes (included with meter kits): One year parts and labor