Please login to your online account to display your discounted pricing
Fisher Scientific™ Accessories for PowerGen™ High Throughput Homogenizers
For use with PowerGen™ High Throughput Homogenizer
$376.80 - $1,703.40
Specifications
For Use With (Equipment) PowerGen™ high throughput homogenizer
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
02-215-506
troemner llc
930165
4mL polyethylene vial with cap and 3/8 in. (9.5mm) stainless-steel bead Case of 240 Each for $1,009.73
02-215-505
troemner llc
930164
4mL polycarbonate vial with cap and 3/8 in. (9.5mm) stainless-steel bead Case of 240 Each for $1,237.82
02-215-507
troemner llc
930166
15mL polycarbonate vial with cap and 7/16 in. (11.1mm) stainless-steel bead Case of 50 Each for $1,371.30
02-215-508
troemner llc
930167
4mL polycarbonate vial with cap only Pack of 240 Each for $627.64
02-215-510
troemner llc
930169
Magnetic pipet tips Pack of 12 Each for $376.80
02-215-512
troemner llc
930171
Stainless-steel grinding beads (0.15 in.) Pack of 5000 Each for $1,261.57
02-215-518
troemner llc
930174
Magnet extractor (24 pin) Each for $1,703.40
02-215-519
troemner llc
930175
Magnet Stand/Ball Separator Each for $553.94
02-215-520
troemner llc
930180
Grinding balls (0.38 in.) Pack of 1000 Each for $1,481.02
Description & Specifications
Specifications
For Use With (Equipment) PowerGen™ high throughput homogenizer
  • Vials are supplied with caps and are available with or without stainless-steel mixing beads
  • Magnetic pipet tips are used to remove beads from microplates
  • Bead dispenser places beads into plates easily and efficiently