Fisherbrand™ Accessories for PowerGen™ High Throughput Homogenizers
For use with PowerGen™ High Throughput Homogenizers.
Manufacturer:  Fisherbrand™ 930169
 View more versions of this product
Catalog No. 02-215-510
Description
Description
  • Vials are supplied with caps and are available with or without stainless-steel mixing beads
  • Magnetic pipet tips are used to remove beads from microplates
  • Bead dispenser places beads into plates easily and efficiently
Specifications
Specifications
Magnetic pipet tips
Ideal for remove beads from microplates
PowerGen™ high throughput homogenizer
Magnet
Documents