Please login to your online account to display your discounted pricing

Fisher Scientific™ Accessories for PowerGen™ High Throughput Homogenizers


For use with PowerGen™ High Throughput Homogenizer

Manufacturer: troemner llc  930165

 View more versions of this product

Catalog No. 02-215-506

  • / Case of 240 Each
 

Description & Specifications

Specifications

For Use With (Equipment) PowerGen™ high throughput homogenizer
Type 4mL polyethylene vial with cap and 3/8 in. (9.5mm) stainless-steel bead
Contains Cap, 0.37 in. stainless steel bead
Capacity (Metric) 4mL
Material Polyethylene