missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Propeller for Overhead Stirrers
Three-bladed Propeller for Overhead Stirrers for general purpose mixing.
Supplier: Fisherbrand™ A521FSR
Description
- Can be used as single or mounted several on a shaft for thorough agitation
- Electro-polished stainless steel 316L
- Secured to shaft with set screws
- Low shear, high top-to-bottom (axial) pumping capacity with high gas injection
- Will vortex 5 gallons of water at 3600 rpm
- High speed, Low to medium viscosity mixing, Axial flow pattern
Specifications
1.5 in. | |
38 mm | |
Electronic mixer | |
4 set screws and allen key | |
Electropolished Stainless Steel |
0.31 in. | |
7.8 mm | |
Ideal for high speed, low to medium viscosity mixing | |
Propeller |