Please login to your online account to display your discounted pricing
Fisher Science Education™ Water Quality pH Pens
Highly stable and accurate readings with large LCD display.
Manufacturer: fisher science education  850050F
 View more versions of this product
Catalog No. S96275C
  • / Each
Description & Specifications
Specifications
Range 2 to 12
For Use With (Application) Field pH tests
Includes Four button cell batteries and a built in electrode kept moist with a protective cap
Accuracy ±0.3
Product Type Water Quality pH Pen